Assay Details
Gene Symbol: Tbc1d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113362.1 | NP_001106833.1 | 8 - 9 | 1514 | 58 | |
NM_001113364.1 | NP_001106835.1 | 5 - 6 | 628 | 58 | ||
NM_133910.3 | NP_598671.3 | 7 - 8 | 1333 | 58 | ||
GenBank mRNA | AB093293.1 | - | 6 - 7 | 1127 | 58 | |
AK013092.1 | - | 5 - 6 | 554 | 58 | ||
AK029986.1 | - | 8 - 9 | 1514 | 58 | ||
BC011420.1 | - | 5 - 6 | 506 | 58 | ||
BC042515.1 | - | 7 - 8 | 1309 | 4020510 | 58 | |
BC096446.1 | - | 7 - 8 | 1325 | 6307706 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
