Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134253.2 | NP_599014.2 | 6 - 7 | 900 | 67 | |
GenBank mRNA | BC005659.1 | - | 6 - 7 | 809 | 3588701 | 67 |
BC006585.1 | - | 2 - 3 | 156 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
