Assay Details
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138607.3 | NP_613073.2 | 12 - 13 | 1189 | 74 | |
GenBank mRNA | AK008402.1 | - | 12 - 13 | 1190 | 74 | |
BC026128.1 | - | 10 - 11 | 1742 | 74 | ||
BC125297.1 | - | 11 - 12 | 1055 | 40129812 | 74 | |
Y18505.1 | - | 12 - 13 | 1154 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467455 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467454 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467453 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467456 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078474 | Y18505.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078474 | NM_138607.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078474 | AK008402.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556085 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971353 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858227 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969147 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426219 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148047 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92514 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159841 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078474 | Y18505.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078474 | NM_138607.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078474 | AK008402.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467450 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467449 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159841 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03698391 | - | - |
Whole Mouse Genome G4122A | A_51_P368557 | BC125297.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P368557 | Y18505.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P368557 | NM_138607.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P368557 | AK008402.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P368557 | BC026128.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107513 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140106 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159841 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14666916 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15153203 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14666918 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14666917 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14958166 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467456 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467454 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467453 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467472 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14590947 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14666917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14925832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15166013 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15166000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15165999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14958166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15128257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467480 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467495 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15128260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14849132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15153204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14782009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32467468 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10600404 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600414 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600408 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600416 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600412 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600413 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600418 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600409 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600405 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600415 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600407 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600411 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600406 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10600410 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419516_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10600403 | - | - |
Mouse Genome 430 2.0 Array | 1419516_at | - | - |
Mouse Genome 430 2.0 Array | 1419516_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108160_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108160_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00004874.mm.1 | TC0X00000710.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00004874.mm.1 | NM_138607.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00004874.mm.1 | NM_138607.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00004873.mm.1 | TC0X00000710.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00004873.mm.1 | NM_138607.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00004873.mm.1 | NM_138607.3 | Refseq |
Mouse Expression Set 430 | 1419516_PM_at | - | - |
Mouse Expression Set 430 | 1419516_at | - | - |
Murine Genome U74v2 Set | 92649_at | - | - |
Murine Genome U74v2 Set | 92649_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419516_at | - | - |
Mouse Genome 430A 2.0 Array | 1419516_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419516_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419516_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 381264 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5670292 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005670292 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687799 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2687799 | - | - |
mouse-6 v1.1 (Illumina) | scl54835.8.1_3-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5670292 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl54835.8.1_3-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670292 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2687799 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2856628 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2856628 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687799 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54835.8.1_3-S | - | - |
