Assay Details
Gene Symbol: Dhx35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291144.1 | NP_001278073.1 | 14 - 15 | 1501 | 70 | |
NM_145742.2 | NP_665685.1 | 14 - 15 | 1481 | 70 | ||
GenBank mRNA | AK004665.1 | - | 14 - 15 | 1427 | 70 | |
AK155482.1 | - | 13 - 14 | 2907 | 70 | ||
AK156955.1 | - | 14 - 15 | 1447 | 70 | ||
AK170952.1 | - | 13 - 14 | 2905 | 70 | ||
BC029709.1 | - | 14 - 15 | 1468 | 5359253 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
