Assay Details
Gene Symbol: Usp45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290425.1 | NP_001277354.1 | 2 - 3 | 157 | 90 | |
NM_152825.2 | NP_690038.1 | 2 - 3 | 157 | 90 | ||
GenBank mRNA | AK019756.1 | - | 2 - 3 | 199 | 90 | |
AK040660.1 | - | 2 - 3 | 158 | 90 | ||
AK088073.1 | - | 2 - 3 | 140 | 90 | ||
AK162825.1 | - | 2 - 3 | 158 | 90 | ||
BB213271.2 | - | 2 - 3 | 169 | 90 | ||
BC027768.1 | - | 2 - 3 | 140 | 5354502 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
