Assay Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019655.3 | NP_062629.3 | 7 - 8 | 2355 | 57 | |
GenBank mRNA | AF052506.1 | - | 7 - 8 | 2268 | 57 | |
AF291050.1 | - | 7 - 8 | 2262 | 57 | ||
AF291875.1 | - | 6 - 7 | 830 | 57 | ||
AK156093.1 | - | 6 - 7 | 922 | 57 | ||
AK172523.1 | - | 6 - 7 | 922 | 57 | ||
BC042505.1 | - | 7 - 8 | 2289 | 4012994 | 57 | |
CT010323.1 | - | 7 - 8 | 2262 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
