Assay Details
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027081.3 | NP_081357.2 | 11 - 12 | 1058 | 102 | |
GenBank mRNA | AK006427.1 | - | 2 - 3 | 195 | 102 | |
BC139003.1 | - | 11 - 12 | 1032 | 8862025 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
