Assay Details
Gene Symbol: Acoxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028765.3 | NP_083041.1 | 7 - 8 | 904 | 67 | |
GenBank mRNA | AK004775.1 | - | 7 - 8 | 906 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
