Assay Details
Gene Symbol: Acoxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028765.3 | NP_083041.1 | 9 - 10 | 1074 | 97 | |
GenBank mRNA | AK004775.1 | - | 9 - 10 | 1076 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
