Assay Details
Gene Symbol: Apeh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146226.2 | NP_666338.2 | 7 - 8 | 780 | 79 | |
GenBank mRNA | AK041271.1 | - | 6 - 7 | 880 | 79 | |
AK149671.1 | - | 7 - 8 | 783 | 79 | ||
AK149713.1 | - | 7 - 8 | 782 | 79 | ||
AK188112.1 | - | 2 - 3 | 198 | 79 | ||
AK212779.1 | - | 2 - 3 | 198 | 79 | ||
AK216818.1 | - | 2 - 3 | 197 | 79 | ||
BC005716.1 | - | 6 - 7 | 906 | 79 | ||
BC025494.1 | - | 7 - 8 | 762 | 5310043 | 79 | |
BC026594.1 | - | 2 - 3 | 313 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
