Assay Details
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026214.2 | NP_001021385.1 | 6 - 7 | 706 | 87 | |
NM_001286049.1 | NP_001272978.1 | 5 - 6 | 681 | 87 | ||
NM_001286058.1 | NP_001272987.1 | 4 - 5 | 608 | 87 | ||
NM_007647.3 | NP_031673.2 | 5 - 6 | 633 | 87 | ||
GenBank mRNA | AF006482.1 | - | 5 - 6 | 641 | 87 | |
AF136571.1 | - | 5 - 6 | 667 | 87 | ||
AJ238636.1 | - | 5 - 6 | 669 | 87 | ||
AK002618.1 | - | 5 - 6 | 606 | 87 | ||
AK031581.1 | - | 6 - 7 | 686 | 87 | ||
AK036641.1 | - | 6 - 7 | 683 | 87 | ||
AK037736.1 | - | 5 - 6 | 602 | 87 | ||
AK045828.1 | - | 5 - 6 | 619 | 87 | ||
AK079267.1 | - | 4 - 5 | 530 | 87 | ||
AK080265.1 | - | 6 - 7 | 693 | 87 | ||
AK081435.1 | - | 6 - 7 | 671 | 87 | ||
AK088455.1 | - | 5 - 6 | 662 | 87 | ||
AK160950.1 | - | 3 - 4 | 527 | 87 | ||
AK163688.1 | - | 5 - 6 | 607 | 87 | ||
BC015247.1 | - | 4 - 5 | 591 | 4242560 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
