Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 5 - 6 | 377 | 67 | |
GenBank mRNA | AK082761.1 | - | 5 - 6 | 378 | 67 | |
AK141692.1 | - | 5 - 6 | 396 | 67 | ||
AK145969.1 | - | 5 - 6 | 386 | 67 | ||
BC034517.1 | - | 4 - 5 | 313 | 4458839 | 67 | |
U97572.1 | - | 4 - 5 | 361 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
