Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 8 - 9 | 735 | 76 | |
GenBank mRNA | AK004652.1 | - | 4 - 5 | 387 | 76 | |
AK082761.1 | - | 8 - 9 | 736 | 76 | ||
AK141692.1 | - | 8 - 9 | 754 | 76 | ||
AK145969.1 | - | 8 - 9 | 744 | 76 | ||
BC034517.1 | - | 7 - 8 | 671 | 4458839 | 76 | |
U97572.1 | - | 7 - 8 | 719 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
