Assay Details
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009602.4 | NP_033732.2 | 3 - 4 | 479 | 65 | |
GenBank mRNA | AF089739.1 | - | 2 - 3 | 169 | 65 | |
AF145286.1 | - | 3 - 4 | 418 | 65 | ||
AF299083.1 | - | 3 - 4 | 273 | 65 | ||
AF325349.1 | - | 3 - 4 | 239 | 65 | ||
AK049722.1 | - | 3 - 4 | 452 | 65 | ||
AK051742.1 | - | 3 - 4 | 461 | 65 | ||
AK081254.1 | - | 456 | 65 | |||
AY574267.1 | - | 3 - 4 | 253 | 65 | ||
BC065103.1 | - | 3 - 4 | 432 | 6838257 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
