Assay Details
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009602.4 | NP_033732.2 | 4 - 5 | 592 | 93 | |
GenBank mRNA | AF089739.1 | - | 3 - 4 | 282 | 93 | |
AF145286.1 | - | 4 - 5 | 531 | 93 | ||
AF299083.1 | - | 4 - 5 | 386 | 93 | ||
AF325349.1 | - | 4 - 5 | 352 | 93 | ||
AK049722.1 | - | 4 - 5 | 565 | 93 | ||
AK051742.1 | - | 4 - 5 | 574 | 93 | ||
AK081254.1 | - | 569 | 93 | |||
AY574267.1 | - | 4 - 5 | 366 | 93 | ||
BC065103.1 | - | 4 - 5 | 545 | 6838257 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
