Assay Details
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010046.2 | NP_034176.1 | 10 - 11 | 967 | 93 | |
GenBank mRNA | AB057816.1 | - | 10 - 11 | 955 | 93 | |
AF078752.1 | - | 10 - 11 | 1025 | 93 | ||
AK215091.1 | - | 4 - 5 | 249 | 93 | ||
BC003717.1 | - | 10 - 11 | 967 | 3486724 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
