Assay Details
Gene Symbol: Gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010266.2 | NP_034396.1 | 1 - 2 | 290 | 97 | |
GenBank mRNA | AF174583.1 | - | 1 - 2 | 216 | 97 | |
AF369880.1 | - | 1 - 2 | 175 | 97 | ||
AF369881.1 | - | 1 - 2 | 175 | 97 | ||
AF369882.1 | - | 1 - 2 | 175 | 97 | ||
AF369883.1 | - | 1 - 2 | 175 | 97 | ||
AF369884.1 | - | 1 - 2 | 175 | 97 | ||
AK041056.1 | - | 1 - 2 | 229 | 97 | ||
AK044078.1 | - | 1 - 2 | 218 | 97 | ||
AK079459.1 | - | 1 - 2 | 230 | 97 | ||
AK157014.1 | - | 1 - 2 | 252 | 97 | ||
AK173138.1 | - | 1 - 2 | 219 | 97 | ||
BC076583.1 | - | 1 - 2 | 203 | 30620457 | 97 | |
BY250819.1 | - | 1 - 2 | 302 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
