Assay Details
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010269.3 | NP_034399.1 | 7 - 8 | 1060 | 71 | |
NR_104341.1 | 7 - 8 | 1022 | 71 | |||
GenBank mRNA | AK004887.1 | - | 7 - 8 | 1006 | 71 | |
AK082958.1 | - | 7 - 8 | 1022 | 71 | ||
AK146835.1 | - | 7 - 8 | 1050 | 71 | ||
AK161122.1 | - | 7 - 8 | 959 | 71 | ||
AK166799.1 | - | 7 - 8 | 975 | 71 | ||
AK166859.1 | - | 7 - 8 | 974 | 71 | ||
BC025070.1 | - | 7 - 8 | 969 | 3710712 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
