Assay Details
Gene Symbol: Gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013530.1 | NP_038558.1 | 9 - 10 | 1300 | 72 | |
GenBank mRNA | AK014789.1 | - | 9 - 10 | 1019 | 72 | |
AK030354.1 | - | 8 - 9 | 1241 | 72 | ||
BC018239.1 | - | 9 - 10 | 1300 | 4511683 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206680 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61768 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100959 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | BC018239.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | AK014789.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | AK030354.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | BC018239.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | AK014789.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817292 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817293 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817294 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817297 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24817250 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100959 | - | - |
Whole Mouse Genome G4122A | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | BC018239.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | NM_013530.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | AK014789.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P293862 | AK030354.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127133 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100959 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05825239 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05825227 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05825241 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05825231 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05499812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24817267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05685583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05685588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05155632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05764165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05685591 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05685589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05155641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05685585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05155645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24817290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05691361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05825241 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10547862 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547860 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547861 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547868 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547865 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547866 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547867 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547863 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10547864 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449159_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10547858 | - | - |
Mouse Genome 430 2.0 Array | 1449159_at | - | - |
Mouse Genome 430 2.0 Array | 1449159_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14695_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14695_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600024285.mm.1 | TC0600003121.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600024285.mm.1 | NM_013530.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600024285.mm.1 | NM_013530.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600024286.mm.1 | TC0600003121.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600024286.mm.1 | NM_013530.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600024286.mm.1 | NM_013530.1 | Refseq |
Mouse Expression Set 430 | 1449159_at | - | - |
Mouse Expression Set 430 | 1449159_PM_at | - | - |
Murine Genome U74v2 Set | 99238_PM_at | - | - |
Murine Genome U74v2 Set | 99238_at | - | - |
Mouse Genome 430A 2.0 Array | 1449159_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449159_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449159_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449159_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 758889 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1230167 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1230167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001230167 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218235 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1218235 | - | - |
mouse-6 v1.1 (Illumina) | scl014695.1_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1230167 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl014695.1_7-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230167 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1218235 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218235 | - | - |
