Assay Details
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013726.3 | NP_038754.1 | 6 - 7 | 797 | 110 | |
GenBank mRNA | AJ003132.1 | - | 6 - 7 | 687 | 110 | |
AK014480.1 | - | 6 - 7 | 763 | 110 | ||
AK083650.1 | - | 3 - 4 | 172 | 110 | ||
AK163408.1 | - | 4 - 5 | 373 | 110 | ||
BC069983.1 | - | 6 - 7 | 745 | 110 | ||
BC138709.1 | - | 6 - 7 | 615 | 8861731 | 110 | |
BC138710.1 | - | 6 - 7 | 615 | 8861732 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
