Assay Details
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 4 - 5 | 666 | 79 | |
GenBank mRNA | AB032602.1 | - | 3 - 4 | 352 | 79 | |
AK052972.1 | - | 5 - 6 | 559 | 79 | ||
BC076594.1 | - | 4 - 5 | 666 | 30649661 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
