Assay Details
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021284.6 | NP_067259.4 | 4 - 5 | 643 | 87 | |
GenBank mRNA | AK155064.1 | - | 4 - 5 | 644 | 87 | |
AK164065.1 | - | 4 - 5 | 644 | 87 | ||
AY487807.1 | - | 3 - 4 | 467 | 87 | ||
BC004642.1 | - | 4 - 5 | 631 | 3158212 | 87 | |
BC010202.1 | - | 4 - 5 | 631 | 3158212 | 87 | |
BC141051.1 | - | 3 - 4 | 469 | 9055663 | 87 | |
X00485.1 | - | 1 - 2 | 127 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
