Assay Details
Gene Symbol: Chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021369.2 | NP_067344.2 | 5 - 6 | 1596 | 71 | |
GenBank mRNA | AJ245706.1 | - | 5 - 6 | 1434 | 71 | |
AK149262.1 | - | 5 - 6 | 1537 | 71 | ||
AY574265.1 | - | 5 - 6 | 1354 | 71 | ||
BC031985.1 | - | 5 - 6 | 1596 | 4501558 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
regulation of dopamine secretion
positive regulation of dopamine secretion
protein heterooligomerization
membrane depolarization
cation transmembrane transport
ion channel activity
extracellular ligand-gated ion channel activity
protein binding
acetylcholine-gated cation-selective channel activity
transport
ion transport
regulation of dopamine secretion
positive regulation of dopamine secretion
protein heterooligomerization
membrane depolarization
cation transmembrane transport
ion channel activity
extracellular ligand-gated ion channel activity
protein binding
acetylcholine-gated cation-selective channel activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041065 | BC031985.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041065 | AK149262.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041065 | NM_021369.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494438 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98466 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138229 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548058 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912198 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351831 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179978 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138736 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041065 | BC031985.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041065 | AK149262.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041065 | NM_021369.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499653 | BC031985.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499653 | AK149262.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499653 | AY574265.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499653 | AJ245706.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499653 | NM_021369.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738145 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738162 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25738171 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138736 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179978 | - | - |
Whole Mouse Genome G4122A | A_51_P499653 | BC031985.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P499653 | AK149262.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P499653 | AY574265.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P499653 | AJ245706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P499653 | NM_021369.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129660 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140820 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138736 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179978 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06898819 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06898812 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06898813 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06892137 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06898817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25738089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06853114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06892141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738173 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07136594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06910151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06853125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07203660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07136587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06892140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07136422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25738159 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10577987 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577982 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577983 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577981 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577989 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577986 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577988 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577985 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10577984 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450426_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450427_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10577980 | - | - |
Mouse Genome 430 2.0 Array | 1450427_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450426_at | - | - |
Mouse Genome 430 2.0 Array | 1450426_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450427_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11440_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11440_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800015557.mm.1 | TC0800001959.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800015557.mm.1 | NM_021369.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800015557.mm.1 | NM_021369.2 | Refseq |
Mouse Expression Set 430 | 1450427_at | - | - |
Mouse Expression Set 430 | 1450426_PM_at | - | - |
Mouse Expression Set 430 | 1450427_PM_at | - | - |
Mouse Expression Set 430 | 1450426_at | - | - |
Murine Genome U74v2 Set | 138177_at | - | - |
Murine Genome U74v2 Set | 138177_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450426_at | - | - |
Mouse Genome 430A 2.0 Array | 1450427_at | - | - |
Mouse Genome 430A 2.0 Array | 1450427_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450426_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450426_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450426_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450427_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450427_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 395875 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5340092 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5340092 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005340092 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732438 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2732438 | - | - |
mouse-6 v1.1 (Illumina) | scl34973.6.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5340092 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34973.6.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340092 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2732438 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2732437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732438 | - | - |
