Assay Details
Gene Symbol: Cdc42ep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021454.3 | NP_067429.1 | 2 - 3 | 340 | 68 | |
GenBank mRNA | AF102773.1 | - | 2 - 3 | 139 | 68 | |
AF164119.1 | - | 2 - 3 | 140 | 68 | ||
AK008154.1 | - | 2 - 3 | 340 | 68 | ||
BC006758.1 | - | 2 - 3 | 166 | 3710012 | 68 | |
BC103774.1 | - | 2 - 3 | 351 | 6529557 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF102773.1,AF164119.1,AK008154.1,BC006758.1,BC103774.1,NM_021454.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948003 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948001 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948002 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057686 | AF102773.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057686 | AF164119.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057686 | AK008154.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057686 | BC006758.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057686 | BC103774.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164637 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184231 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507810 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427826 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10888 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945408 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 59491 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057686 | AF102773.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057686 | AF164119.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057686 | AK008154.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057686 | BC006758.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057686 | BC103774.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286018 | AF102773.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286018 | AF164119.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286018 | AK008154.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286018 | BC006758.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P286018 | BC103774.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165679 | AF164119.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165679 | AK008154.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165679 | BC006758.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165679 | BC103774.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P165679 | NM_021454.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948042 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948059 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948061 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948068 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948054 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948010 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24948066 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06113207 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06113218 | - | - |
Whole Mouse Genome G4122A | A_52_P40191 | - | - |
Whole Mouse Genome G4122A | A_51_P286018 | AF102773.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286018 | AF164119.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286018 | AK008154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286018 | BC006758.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P286018 | BC103774.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165679 | AF164119.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165679 | AK008154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165679 | BC006758.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165679 | BC103774.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P165679 | NM_021454.3 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06064152 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06064151 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05947033 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06064154 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05947035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948002 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948000 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948003 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24948001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24948007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24947946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06555421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06296065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05984646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06041789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06013964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06070613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06013962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06280493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06313073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24947999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06313072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06013963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24948007 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10559512 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559510 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418712_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430018_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10559509 | - | - |
Mouse Genome 430 2.0 Array | 1430018_at | - | - |
Mouse Genome 430 2.0 Array | 1430018_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418712_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418712_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58804_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58804_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700019524.mm.1 | TC0700002148.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700019524.mm.1 | NM_021454.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019524.mm.1 | NM_021454.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019523.mm.1 | TC0700002148.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700019523.mm.1 | NM_021454.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019523.mm.1 | NM_021454.3 | Refseq |
Mouse Expression Set 430 | 1418712_at | - | - |
Mouse Expression Set 430 | 1418712_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418712_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418712_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430018_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418712_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1430018_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418712_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 917585 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6400397 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6400397 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006400397 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592823 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2546494 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2592823 | - | - |
mouse-6 v1.1 (Illumina) | scl31823.4.1_170-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6400397 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl31823.4.1_170-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400397 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2592823 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592823 | - | - |
