Assay Details
Gene Symbol: Cnot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037846.3 | NP_001032935.2 | 10 - 11 | 1176 | 116 | |
NM_001037847.2 | NP_001032936.2 | 10 - 11 | 1106 | 116 | ||
NM_028082.2 | NP_082358.2 | 9 - 10 | 1053 | 116 | ||
NR_024327.1 | 8 - 9 | 986 | 116 | |||
GenBank mRNA | AK011231.1 | - | 8 - 9 | 976 | 116 | |
AK078121.1 | - | 10 - 11 | 1166 | 116 | ||
AK149767.1 | - | 10 - 11 | 1150 | 116 | ||
AK167834.1 | - | 10 - 11 | 1154 | 116 | ||
BC043133.1 | - | 10 - 11 | 1077 | 116 | ||
BC063105.1 | - | 11 - 12 | 1319 | 5003026 | 116 | |
BC065171.1 | - | 10 - 11 | 1106 | 6415034 | 116 | |
BC090624.1 | - | 8 - 9 | 982 | 6815664 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
