Assay Details
Gene Symbol: Slc8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177594.1 | NP_001171065.1 | 1 - 2 | 92 | 91 | |
NM_001177595.1 | NP_001171066.1 | 1 - 2 | 92 | 91 | ||
NM_133221.2 | NP_573484.2 | 1 - 2 | 92 | 91 | ||
GenBank mRNA | AF261233.1 | - | 1 - 2 | 92 | 91 | |
AK028537.1 | - | 1 - 2 | 37 | 91 | ||
AK157173.1 | - | 1 - 2 | 50 | 91 | ||
BC043689.1 | - | 1 - 2 | 32 | 5137181 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF261233.1,AK157173.1,BC043689.1,NM_001177594.1,NM_001177595.1,NM_133221.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
