Assay Details
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133973.2 | NP_598734.1 | 13 - 14 | 1729 | 73 | |
GenBank mRNA | AK156466.1 | - | 13 - 14 | 1721 | 73 | |
AK172465.1 | - | 13 - 14 | 1720 | 73 | ||
AK188268.1 | - | 3 - 4 | 315 | 73 | ||
AK188304.1 | - | 3 - 4 | 313 | 73 | ||
AY223866.1 | - | 13 - 14 | 1756 | 73 | ||
BC023120.1 | - | 13 - 14 | 1706 | 4161858 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
