Assay Details
Gene Symbol: Aldh7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127338.1 | NP_001120810.1 | 1 - 2 | 99 | 58 | |
GenBank mRNA | AK004991.1 | - | 1 - 2 | 33 | 58 | |
CJ156580.1 | - | 1 - 2 | 99 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004991.1, NM_001127338.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
