Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF402611.1 | - | 8 - 9 | 1454 | 91 | |
AF402612.1 | - | 7 - 8 | 1351 | 91 | ||
AF402613.1 | - | 8 - 9 | 1501 | 91 | ||
AK028302.1 | - | 5 - 6 | 903 | 91 | ||
AK034504.1 | - | 7 - 8 | 1444 | 91 | ||
AK054242.1 | - | 8 - 9 | 1442 | 91 | ||
AK134961.1 | - | 8 - 9 | 1517 | 91 | ||
AK149626.1 | - | 8 - 9 | 1505 | 91 | ||
BC129902.1 | - | 8 - 9 | 1541 | 40111637 | 91 | |
BC129903.1 | - | 8 - 9 | 1542 | 40111639 | 91 | |
BC145523.1 | - | 8 - 9 | 1595 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
