Assay Details
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139272.2 | NP_644678.2 | 14 - 15 | 1572 | 82 | |
GenBank mRNA | AF348968.1 | - | 14 - 15 | 1440 | 82 | |
AK046567.1 | - | 14 - 15 | 1530 | 82 | ||
AK145122.1 | - | 14 - 15 | 1450 | 82 | ||
AK149609.1 | - | 14 - 15 | 1500 | 82 | ||
AK151536.1 | - | 14 - 15 | 1363 | 82 | ||
BC007172.1 | - | 13 - 14 | 1307 | 82 | ||
BC053063.1 | - | 13 - 14 | 1415 | 5702572 | 82 | |
BC059818.1 | - | 14 - 15 | 1517 | 5697789 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
