Assay Details
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144553.2 | NP_653136.2 | 16 - 17 | 2349 | 91 | |
GenBank mRNA | AB076696.1 | - | 16 - 17 | 2349 | 91 | |
AK036384.1 | - | 15 - 16 | 3587 | 91 | ||
AK048470.1 | - | 16 - 17 | 2120 | 91 | ||
AK085882.1 | - | 16 - 17 | 2130 | 91 | ||
AK129032.1 | - | 16 - 17 | 2095 | 91 | ||
AK135820.1 | - | 16 - 17 | 2156 | 91 | ||
AK147955.1 | - | 16 - 17 | 2119 | 91 | ||
BC027273.1 | - | 16 - 17 | 2070 | 91 | ||
BC043924.1 | - | 8 - 9 | 975 | 91 | ||
BC058087.1 | - | 16 - 17 | 2083 | 6816660 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
