Assay Details
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145129.2 | NP_660111.2 | 3 - 4 | 405 | 88 | |
GenBank mRNA | AF325346.1 | - | 3 - 4 | 275 | 88 | |
AF459029.1 | - | 3 - 4 | 378 | 88 | ||
AF472588.1 | - | 3 - 4 | 307 | 88 | ||
AK051730.1 | - | 3 - 4 | 409 | 88 | ||
AK053497.1 | - | 3 - 4 | 333 | 88 | ||
AK080415.1 | - | 3 - 4 | 406 | 88 | ||
AK080475.1 | - | 3 - 4 | 406 | 88 | ||
AK148786.1 | - | 3 - 4 | 377 | 88 | ||
AY574262.1 | - | 3 - 4 | 240 | 88 | ||
BC120597.1 | - | 3 - 4 | 276 | 40129520 | 88 | |
BC120599.1 | - | 3 - 4 | 276 | 40129522 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
