Assay Details
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145130.2 | NP_660112.1 | 2 - 3 | 366 | 62 | |
GenBank mRNA | AB294194.1 | - | 2 - 3 | 261 | 62 | |
AK098090.1 | - | 2 - 3 | 337 | 62 | ||
AK154062.1 | - | 2 - 3 | 361 | 62 | ||
AK165319.1 | - | 2 - 3 | 344 | 62 | ||
AK169452.1 | - | 2 - 3 | 343 | 62 | ||
AY028317.1 | - | 2 - 3 | 342 | 62 | ||
BC006753.1 | - | 2 - 3 | 294 | 3709076 | 62 | |
CN705185.1 | - | 2 - 3 | 366 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
