Assay Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285988.1 | NP_001272917.1 | 9 - 10 | 1318 | 109 | |
NM_001285989.1 | NP_001272918.1 | 9 - 10 | 1300 | 109 | ||
NM_001285990.1 | NP_001272919.1 | 8 - 9 | 1220 | 109 | ||
NM_145436.2 | NP_663411.2 | 9 - 10 | 1300 | 109 | ||
GenBank mRNA | AK039377.1 | - | 9 - 10 | 1097 | 109 | |
AK134425.1 | - | 8 - 9 | 1012 | 109 | ||
AK148278.1 | - | 9 - 10 | 1155 | 109 | ||
AK161471.1 | - | 9 - 10 | 1169 | 109 | ||
BC023187.1 | - | 9 - 10 | 1176 | 109 | ||
BC157955.1 | - | 9 - 10 | 1317 | 9088128 | 109 | |
BC172100.1 | - | 9 - 10 | 1317 | 9054386 | 109 | |
BC172149.1 | - | 8 - 9 | 1219 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
