Assay Details
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK145200.1 | - | 18 - 19 | 1957 | 96 | |
AK161473.1 | - | 18 - 19 | 2040 | 96 | ||
BC030328.1 | - | 18 - 19 | 1957 | 5376020 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
