Assay Details
Gene Symbol: Tmem63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144794.2 | NP_659043.1 | 15 - 16 | 1538 | 81 | |
GenBank mRNA | AK049360.1 | - | 15 - 16 | 1533 | 81 | |
AK144899.1 | - | 15 - 16 | 1591 | 81 | ||
AK170559.1 | - | 15 - 16 | 1554 | 81 | ||
AK173027.1 | - | 15 - 16 | 1531 | 81 | ||
BC014795.1 | - | 15 - 16 | 1511 | 3961992 | 81 | |
BC019442.1 | - | 15 - 16 | 1538 | 4036849 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
