Assay Details
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144813.1 | NP_659062.1 | 1 - 2 | 125 | 106 | |
GenBank mRNA | BC016094.1 | - | 1 - 2 | 125 | 4504496 | 106 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC016094.1, NM_144813.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
