Assay Details
Gene Symbol: Mtmr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144843.4 | NP_659092.1 | 7 - 8 | 988 | 66 | |
GenBank mRNA | AB517623.1 | - | 8 - 9 | 969 | 66 | |
AK076218.1 | - | 7 - 8 | 965 | 66 | ||
AK154859.1 | - | 7 - 8 | 908 | 66 | ||
BC020019.1 | - | 7 - 8 | 922 | 3596732 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
