Assay Details
Gene Symbol: Eml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144872.1 | NP_659121.1 | 10 - 11 | 1438 | 86 | |
GenBank mRNA | BC022146.1 | - | 10 - 11 | 1438 | 5101626 | 86 |
BC023878.1 | - | 10 - 11 | 1461 | 5354867 | 86 | |
BC034112.1 | - | 10 - 11 | 1236 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
