Assay Details
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177445.5 | NP_803228.2 | 5 - 6 | 622 | 67 | |
GenBank mRNA | AK077533.1 | - | 5 - 6 | 545 | 67 | |
AK144470.1 | - | 5 - 6 | 567 | 67 | ||
AK146350.1 | - | 5 - 6 | 566 | 67 | ||
AK151027.1 | - | 5 - 6 | 566 | 67 | ||
AK151897.1 | - | 5 - 6 | 533 | 67 | ||
AK152304.1 | - | 5 - 6 | 533 | 67 | ||
AK152562.1 | - | 5 - 6 | 533 | 67 | ||
AK153256.1 | - | 5 - 6 | 533 | 67 | ||
AK153299.1 | - | 5 - 6 | 533 | 67 | ||
AK166009.1 | - | 5 - 6 | 533 | 67 | ||
AK169247.1 | - | 5 - 6 | 533 | 67 | ||
AK169716.1 | - | 5 - 6 | 510 | 67 | ||
BC008638.1 | - | 5 - 6 | 520 | 3586278 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
