Assay Details
Gene Symbol: Cwh43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181323.2 | NP_851840.1 | 9 - 10 | 1549 | 96 | |
GenBank mRNA | AK035550.1 | - | 8 - 9 | 1142 | 96 | |
AK081976.1 | - | 9 - 10 | 1549 | 96 | ||
BC016523.1 | - | 9 - 10 | 3584006 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
