Assay Details
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145970.1 | NP_666082.2 | 16 - 17 | 1851 | 63 | |
GenBank mRNA | BC016188.1 | - | 16 - 17 | 1851 | 4482793 | 63 |
BC027028.1 | - | 15 - 16 | 1666 | 5363547 | 63 | |
DQ329239.1 | - | 9 - 10 | 901 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
