Assay Details
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285785.1 | NP_001272714.1 | 6 - 7 | 1006 | 73 | |
NM_146011.2 | NP_666123.2 | 5 - 6 | 445 | 73 | ||
GenBank mRNA | AK047280.1 | - | 5 - 6 | 390 | 73 | |
AK049245.1 | - | 2 - 3 | 226 | 73 | ||
AK080955.1 | - | 4 - 5 | 1140 | 73 | ||
AK083474.1 | - | 2 - 3 | 355 | 73 | ||
AK089051.1 | - | 5 - 6 | 397 | 73 | ||
AK167738.1 | - | 6 - 7 | 583 | 73 | ||
BC024535.1 | - | 5 - 6 | 440 | 4987970 | 73 | |
BC027374.1 | - | 5 - 6 | 416 | 4947306 | 73 | |
DQ498128.1 | - | 6 - 7 | 854 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK047280.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
