Assay Details
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146065.1 | NP_666177.1 | 2 - 3 | 128 | 71 | |
GenBank mRNA | AK039648.1 | - | 119 | 71 | ||
AK047901.1 | - | 104 | 71 | |||
AK154588.1 | - | 119 | 71 | |||
AK155046.1 | - | 2 - 3 | 119 | 71 | ||
AK155444.1 | - | 2 - 3 | 146 | 71 | ||
AK157628.1 | - | 158 | 71 | |||
BC026483.1 | - | 2 - 3 | 128 | 4503463 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
