Assay Details
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 18 - 19 | 2244 | 69 | |
NM_146157.4 | NP_666269.2 | 19 - 20 | 2253 | 69 | ||
GenBank mRNA | AF525925.1 | - | 9 - 10 | 1092 | 69 | |
AK049061.1 | - | 19 - 20 | 2244 | 69 | ||
AK077496.1 | - | 19 - 20 | 2244 | 69 | ||
AK089289.1 | - | 18 - 19 | 2223 | 69 | ||
AK129052.1 | - | 19 - 20 | 2206 | 69 | ||
AK161677.1 | - | 18 - 19 | 2209 | 69 | ||
AK170739.1 | - | 18 - 19 | 2223 | 69 | ||
BC056952.1 | - | 14 - 15 | 1973 | 69 | ||
BC129804.1 | - | 19 - 20 | 2240 | 40087024 | 69 | |
BC139366.1 | - | 19 - 20 | 2239 | 8862388 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
