Assay Details
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK150411.1 | - | 1 - 2 | 61 | 82 | |
AK155912.1 | - | 1 - 2 | 79 | 82 | ||
AK170019.1 | - | 1 - 2 | 84 | 82 | ||
X13335.1 | - | 1 - 2 | 58 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
