Assay Details
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027886.3 | NP_082162.3 | 4 - 5 | 483 | 85 | |
GenBank mRNA | AK004757.2 | - | 4 - 5 | 436 | 85 | |
AK156122.1 | - | 3 - 4 | 571 | 85 | ||
AK165911.1 | - | 4 - 5 | 474 | 85 | ||
AK171989.1 | - | 3 - 4 | 345 | 85 | ||
AK210261.1 | - | 2 - 3 | 197 | 85 | ||
BC042403.1 | - | 3 - 4 | 342 | 85 | ||
BC061635.1 | - | 4 - 5 | 466 | 6530178 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
