Assay Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172146.2 | NP_742158.1 | 1 - 2 | 622 | 82 | |
GenBank mRNA | AK147750.1 | - | 1 - 2 | 272 | 82 | |
AK148034.1 | - | 1 - 2 | 226 | 82 | ||
AK160542.1 | - | 1 - 2 | 247 | 82 | ||
AK164506.1 | - | 1 - 2 | 236 | 82 | ||
AK167057.1 | - | 1 - 2 | 261 | 82 | ||
BC006053.1 | - | 1 - 2 | 209 | 82 | ||
BC023841.1 | - | 1 - 2 | 249 | 5346105 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
