Assay Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172146.2 | NP_742158.1 | 4 - 5 | 1007 | 79 | |
GenBank mRNA | AK147750.1 | - | 4 - 5 | 657 | 79 | |
AK148034.1 | - | 4 - 5 | 611 | 79 | ||
AK159141.1 | - | 5 - 6 | 1157 | 79 | ||
AK159859.1 | - | 5 - 6 | 1157 | 79 | ||
AK160542.1 | - | 4 - 5 | 632 | 79 | ||
AK164506.1 | - | 4 - 5 | 621 | 79 | ||
AK167057.1 | - | 4 - 5 | 646 | 79 | ||
BC006053.1 | - | 4 - 5 | 594 | 79 | ||
BC023841.1 | - | 4 - 5 | 634 | 5346105 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
