Assay Details
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172410.2 | NP_765998.1 | 7 - 8 | 662 | 79 | |
GenBank mRNA | AK010450.1 | - | 6 - 7 | 613 | 79 | |
AK028864.1 | - | 7 - 8 | 663 | 79 | ||
AK045380.1 | - | 521 | 79 | |||
AK129055.1 | - | 6 - 7 | 612 | 79 | ||
AK167105.1 | - | 6 - 7 | 613 | 79 | ||
AK169194.1 | - | 4 - 5 | 383 | 79 | ||
AK169693.1 | - | 6 - 7 | 632 | 79 | ||
BC023140.1 | - | 6 - 7 | 608 | 3991335 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
